Assay Details
| Gene Symbol: TM9SF2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039296.1 | NP_001034385.1 | 5 - 6 | 630 | 127 | |
| GenBank mRNA | AJ719709.1 | - | 5 - 6 | 630 | 127 | |
More Information
Set Membership: |
|

TM9SF2L
Genomic Map