Assay Details
| Gene Symbol: CLTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039313.1 | NP_001034402.1 | 2 - 3 | 315 | 67 | |
| XM_015280417.2 | XP_015135903.1 | 2 - 3 | 326 | 67 | ||
| XM_015280418.2 | XP_015135904.1 | 2 - 3 | 326 | 67 | ||
| GenBank mRNA | AJ721067.1 | - | 2 - 3 | 315 | 67 | |
More Information
Set Membership: |
|

CLTA
Genomic Map