Assay Details
| Gene Symbol: RARRES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204534.4 | NP_989865.1 | 1 - 2 | 253 | 69 | |
| GenBank mRNA | AJ307060.2 | - | 1 - 2 | 253 | 69 | |
More Information
Set Membership: |
|

RARRES1
Genomic Map