Assay Details
| Gene Symbol: RDM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204546.2 | NP_989877.2 | 3 - 4 | 403 | 81 | |
| XM_015299306.2 | XP_015154792.1 | 3 - 4 | 402 | 81 | ||
| XM_015299307.2 | XP_015154793.1 | 3 - 4 | 411 | 81 | ||
| GenBank mRNA | AB080727.1 | - | 3 - 4 | 403 | 81 | |
| CB016342.1 | - | 3 - 4 | 378 | 81 | ||
More Information
Set Membership: |
|

RDM1
Genomic Map