Assay Details
| Gene Symbol: RPS19BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006239.3 | NP_001006239.2 | 3 - 4 | 324 | 62 | |
| GenBank mRNA | AJ719419.1 | - | 3 - 4 | 292 | 62 | |
| CR353921.1 | - | 3 - 4 | 324 | 62 | ||
More Information
Set Membership: |
|

RPS19BP1
Genomic Map