Assay Details
| Gene Symbol: HNRNPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006309.3 | NP_001006309.3 | 6 - 7 | 792 | 57 | |
| XM_015297543.2 | XP_015153029.1 | 6 - 7 | 842 | 57 | ||
| XM_015297545.2 | XP_015153031.1 | 4 - 5 | 542 | 57 | ||
| XM_015297546.2 | XP_015153032.1 | 5 - 6 | 612 | 57 | ||
| XM_025142983.1 | XP_024998751.1 | 6 - 7 | 783 | 57 | ||
| GenBank mRNA | AJ446673.1 | - | 1 - 2 | 174 | 57 | |
| AJ720877.1 | - | 6 - 7 | 787 | 57 | ||
| CN779270.1 | - | 3 - 4 | 403 | 57 | ||
| CV038982.1 | - | 1 - 2 | 40 | 57 | ||
More Information
Set Membership: |
|

HNRNPR
Genomic Map