Assay Details
| Gene Symbol: ARHGAP11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006479.2 | NP_001006479.2 | 5 - 6 | 770 | 107 | |
| XM_015287342.2 | XP_015142828.1 | 5 - 6 | 817 | 107 | ||
| XM_015287343.2 | XP_015142829.1 | 5 - 6 | 671 | 107 | ||
| XM_025150725.1 | XP_025006493.1 | 7 - 8 | 5844 | 107 | ||
| GenBank mRNA | AY313784.1 | - | 5 - 6 | 770 | 107 | |
More Information
Set Membership: |
|

ARHGAP11B
Genomic Map