Assay Details
| Gene Symbol: ARHGAP11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006479.2 | NP_001006479.2 | 6 - 7 | 908 | 67 | |
| GenBank mRNA | AY313784.1 | - | 6 - 7 | 908 | 67 | |
More Information
Set Membership: |
|

ARHGAP11B
Genomic Map