Assay Details
| Gene Symbol: OCM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007477.3 | NP_001007478.1 | 2 - 3 | 124 | 99 | |
| GenBank mRNA | BU221002.1 | - | 2 - 3 | 109 | 99 | |
| BU442436.1 | - | 2 - 3 | 118 | 99 | ||
| CO505144.1 | - | 2 - 3 | 193 | 99 | ||
| M94894.1 | - | 1 - 2 | 67 | 99 | ||
More Information
Set Membership: |
|

OCM3
Genomic Map