Assay Details
| Gene Symbol: OCM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007477.3 | NP_001007478.1 | 3 - 4 | 252 | 69 | |
| GenBank mRNA | BU221002.1 | - | 3 - 4 | 237 | 69 | |
| BU442436.1 | - | 3 - 4 | 246 | 69 | ||
| CO505144.1 | - | 3 - 4 | 321 | 69 | ||
| M34330.1 | - | 1 - 2 | 75 | 69 | ||
| M94894.1 | - | 2 - 3 | 195 | 69 | ||
More Information
Set Membership: |
|

OCM3
Genomic Map