Assay Details
| Gene Symbol: OCM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007477.3 | NP_001007478.1 | 4 - 5 | 357 | 58 | |
| GenBank mRNA | BU221002.1 | - | 4 - 5 | 342 | 58 | |
| BU442436.1 | - | 4 - 5 | 351 | 58 | ||
| CO505144.1 | - | 4 - 5 | 426 | 58 | ||
More Information
Set Membership: |
|

OCM3
Genomic Map