Assay Details
| Gene Symbol: TLR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011691.3 | NP_001011691.3 | 3 - 4 | 2460 | 116 | |
| XM_025149682.1 | XP_025005450.1 | 4 - 5 | 2523 | 116 | ||
| GenBank mRNA | DQ780341.2 | - | 3 - 4 | 2460 | 116 | |
| JF273967.1 | - | 5 - 6 | 2298 | 116 | ||
| KM486590.1 | - | 1 - 2 | 1767 | 116 | ||
| KM486591.1 | - | 1 - 2 | 1767 | 116 | ||
| KM486593.1 | - | 3 - 4 | 2271 | 116 | ||
| KM486594.1 | - | 3 - 4 | 2271 | 116 | ||
| KM486595.1 | - | 3 - 4 | 2271 | 116 | ||
| KM486596.1 | - | 3 - 4 | 2271 | 116 | ||
| KM486597.1 | - | 3 - 4 | 2271 | 116 | ||
| KM486598.1 | - | 1 - 2 | 1767 | 116 | ||
More Information
Set Membership: |
|

TLR3
Genomic Map