Assay Details
| Gene Symbol: EXOSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012798.2 | NP_001012816.2 | 3 - 4 | 304 | 69 | |
| GenBank mRNA | AJ721096.1 | - | 303 | 69 | ||
| BM440764.1 | - | 3 - 4 | 276 | 69 | ||
| BX930470.1 | - | 3 - 4 | 267 | 69 | ||
| CR352497.1 | - | 3 - 4 | 256 | 69 | ||
More Information
Set Membership: |
|

EXOSC2
Genomic Map