Assay Details
| Gene Symbol: SELENOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024734.3 | NP_001019905.1 | 1 - 2 | 121 | 65 | |
| GenBank mRNA | AL588177.1 | - | 1 - 2 | 122 | 65 | |
| BX931291.2 | - | 1 - 2 | 84 | 65 | ||
| BX934085.2 | - | 1 - 2 | 109 | 65 | ||
More Information
Set Membership: |
|

SELENOS
Genomic Map