Assay Details
| Gene Symbol: CDC2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030871.3 | NP_001026042.2 | 12 - 13 | 1432 | 145 | |
| XM_015296863.2 | XP_015152349.1 | 13 - 14 | 4282 | 145 | ||
| XM_015296864.2 | XP_015152350.1 | 13 - 14 | 4279 | 145 | ||
| XM_015296865.2 | XP_015152351.1 | 13 - 14 | 4276 | 145 | ||
| XM_015296866.2 | XP_015152352.1 | 12 - 13 | 4159 | 145 | ||
| XM_025142506.1 | XP_024998274.1 | 13 - 14 | 4300 | 145 | ||
| XM_025142508.1 | XP_024998276.1 | 12 - 13 | 4156 | 145 | ||
More Information
Set Membership: |
|

CDC2L1
Genomic Map