Assay Details
| Gene Symbol: CDC2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030871.3 | NP_001026042.2 | 13 - 14 | 1537 | 82 | |
| XM_015296863.2 | XP_015152349.1 | 14 - 15 | 4387 | 82 | ||
| XM_015296864.2 | XP_015152350.1 | 14 - 15 | 4384 | 82 | ||
| XM_015296865.2 | XP_015152351.1 | 14 - 15 | 4381 | 82 | ||
| XM_015296866.2 | XP_015152352.1 | 13 - 14 | 4264 | 82 | ||
| XM_025142506.1 | XP_024998274.1 | 14 - 15 | 4405 | 82 | ||
| XM_025142508.1 | XP_024998276.1 | 13 - 14 | 4261 | 82 | ||
More Information
Set Membership: |
|

CDC2L1
Genomic Map