Assay Details
| Gene Symbol: CDC2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030871.3 | NP_001026042.2 | 2 - 3 | 225 | 90 | |
| XM_015296863.2 | XP_015152349.1 | 3 - 4 | 3069 | 90 | ||
| XM_015296864.2 | XP_015152350.1 | 3 - 4 | 3069 | 90 | ||
| XM_015296865.2 | XP_015152351.1 | 3 - 4 | 3069 | 90 | ||
| XM_015296866.2 | XP_015152352.1 | 3 - 4 | 3069 | 90 | ||
| XM_025142506.1 | XP_024998274.1 | 3 - 4 | 3087 | 90 | ||
| XM_025142508.1 | XP_024998276.1 | 3 - 4 | 3069 | 90 | ||
| GenBank mRNA | CR390730.1 | - | 3 - 4 | 327 | 90 | |
More Information
Set Membership: |
|

CDC2L1
Genomic Map