Assay Details
| Gene Symbol: CDC2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030871.3 | NP_001026042.2 | 9 - 10 | 1049 | 82 | |
| XM_015296865.2 | XP_015152351.1 | 10 - 11 | 3893 | 82 | ||
More Information
Set Membership: |
|

CDC2L1
Genomic Map