Assay Details
| Gene Symbol: BET1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031173.2 | NP_001026344.2 | 3 - 4 | 179 | 65 | |
| GenBank mRNA | AJ719590.1 | - | 3 - 4 | 179 | 65 | |
More Information
Set Membership: |
|

BET1L
Genomic Map