Assay Details
| Gene Symbol: SELENOP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031609.2 | NP_001026780.2 | 3 - 4 | 471 | 92 | |
| GenBank mRNA | AJ719438.1 | - | 3 - 4 | 471 | 92 | |
More Information
Set Membership: |
|

SELENOP1
Genomic Map