Assay Details
| Gene Symbol: LOC395100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039596.1 | NP_001034685.1 | 826 | 88 | ||
| XM_015298681.2 | XP_015154167.1 | 975 | 88 | |||
| XM_015298682.2 | XP_015154168.1 | 1995 | 88 | |||
| GenBank mRNA | AY300013.1 | - | 826 | 88 | ||
More Information
Set Membership: |
|

LOC395100
Genomic Map