Assay Details
| Gene Symbol: COLEC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039599.1 | NP_001034688.1 | 8 - 9 | 2121 | 73 | |
| GenBank mRNA | DQ157755.1 | - | 8 - 9 | 2121 | 73 | |
More Information
Set Membership: |
|

COLEC12
Genomic Map