Assay Details
| Gene Symbol: PRKAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039605.1 | NP_001034694.1 | 6 - 7 | 830 | 62 | |
| XM_015291061.2 | XP_015146547.1 | 6 - 7 | 62 | |||
| GenBank mRNA | DQ340396.1 | - | 6 - 7 | 830 | 62 | |
More Information
Set Membership: |
|

PRKAA2
Genomic Map