Assay Details
| Gene Symbol: PRKAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039912.1 | NP_001035001.1 | 3 - 4 | 447 | 65 | |
| XM_015275334.2 | XP_015130820.2 | 3 - 4 | 3720 | 65 | ||
| XM_015275335.2 | XP_015130821.1 | 3 - 4 | 442 | 65 | ||
| GenBank mRNA | BX936039.2 | - | 1020 | 65 | ||
| DQ398947.1 | - | 3 - 4 | 447 | 65 | ||
More Information
Set Membership: |
|

PRKAB1
Genomic Map