Assay Details
| Gene Symbol: OLFM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040463.1 | NP_001035553.1 | 2 - 3 | 155 | 73 | |
| GenBank mRNA | AB236174.1 | - | 2 - 3 | 155 | 73 | |
More Information
Set Membership: |
|

OLFM4
Genomic Map