Assay Details
| Gene Symbol: CENPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044635.1 | NP_001038100.1 | 5 - 6 | 573 | 89 | |
| XM_015293048.2 | XP_015148534.1 | 6 - 7 | 763 | 89 | ||
| XM_015293049.2 | XP_015148535.1 | 6 - 7 | 829 | 89 | ||
| XM_015293050.2 | XP_015148536.1 | 6 - 7 | 845 | 89 | ||
| XM_015293051.2 | XP_015148537.1 | 6 - 7 | 727 | 89 | ||
| GenBank mRNA | AB231851.1 | - | 5 - 6 | 573 | 89 | |
| BX932979.1 | - | 6 - 7 | 732 | 89 | ||
More Information
Set Membership: |
|

CENPP
Genomic Map