Assay Details
Gene Symbol: PARD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044641.2 | NP_001038106.1 | 1 - 2 | 262 | 92 | |
GenBank mRNA | BX930407.1 | - | 1 - 2 | 225 | 92 | |
BX932654.1 | - | 1 - 2 | 225 | 92 | ||
DQ683188.1 | - | 1 - 2 | 262 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
