Assay Details
| Gene Symbol: PIGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044644.1 | NP_001038109.1 | 2057 | 56 | ||
| XM_015298928.1 | XP_015154414.1 | 2052 | 56 | |||
| GenBank mRNA | AF303371.1 | - | 1816 | 56 | ||
More Information
Set Membership: |
|

PIGR
Genomic Map