Assay Details
| Gene Symbol: NSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044646.1 | NP_001038111.1 | 4 - 5 | 526 | 73 | |
| XM_025148669.1 | XP_025004437.1 | 4 - 5 | 391 | 73 | ||
| GenBank mRNA | AB253767.1 | - | 4 - 5 | 526 | 73 | |
More Information
Set Membership: |
|

NSL1
Genomic Map