Assay Details
| Gene Symbol: CENPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044649.1 | NP_001038114.1 | 2 - 3 | 208 | 79 | |
| XM_015285134.2 | XP_015140620.1 | 3 - 4 | 312 | 79 | ||
| XM_015285135.2 | XP_015140621.1 | 3 - 4 | 345 | 79 | ||
| XM_015285136.2 | XP_015140622.1 | 3 - 4 | 312 | 79 | ||
| GenBank mRNA | AB253770.1 | - | 2 - 3 | 208 | 79 | |
| BX931524.1 | - | 3 - 4 | 263 | 79 | ||
More Information
Set Membership: |
|

CENPQ
Genomic Map