Assay Details
| Gene Symbol: PACSIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044652.1 | NP_001038117.1 | 4 - 5 | 412 | 67 | |
| XM_015287027.2 | XP_015142513.1 | 4 - 5 | 376 | 67 | ||
| XM_025150708.1 | XP_025006476.1 | 3 - 4 | 354 | 67 | ||
| GenBank mRNA | BX935820.2 | - | 2 - 3 | 245 | 67 | |
| BX935888.2 | - | 4 - 5 | 412 | 67 | ||
| DQ508438.1 | - | 4 - 5 | 412 | 67 | ||
More Information
Set Membership: |
|

PACSIN3
Genomic Map