Assay Details
| Gene Symbol: PTGFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044657.2 | NP_001038122.1 | 1 - 2 | 853 | 135 | |
| XM_015290822.2 | XP_015146308.1 | 2 - 3 | 916 | 135 | ||
| XM_015290823.2 | XP_015146309.1 | 2 - 3 | 1034 | 135 | ||
| XM_015290824.2 | XP_015146310.1 | 2 - 3 | 1015 | 135 | ||
| XM_025152864.1 | XP_025008632.1 | 3 - 4 | 1644 | 135 | ||
| XM_025152865.1 | XP_025008633.1 | 2 - 3 | 1323 | 135 | ||
| XM_025152866.1 | XP_025008634.1 | 2 - 3 | 1348 | 135 | ||
| GenBank mRNA | DQ200257.1 | - | 1 - 2 | 854 | 135 | |
More Information
Set Membership: |
|

PTGFR
Genomic Map