Assay Details
| Gene Symbol: PRKAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044662.1 | NP_001038127.1 | 2 - 3 | 628 | 68 | |
| XM_015290310.2 | XP_015145796.1 | 3 - 4 | 385 | 68 | ||
| XM_015290311.2 | XP_015145797.1 | 3 - 4 | 508 | 68 | ||
| XM_015290312.2 | XP_015145798.1 | 2 - 3 | 513 | 68 | ||
| XM_015290313.2 | XP_015145799.1 | 3 - 4 | 385 | 68 | ||
| XM_015290314.2 | XP_015145800.1 | 3 - 4 | 447 | 68 | ||
| XM_015290315.2 | XP_015145801.1 | 3 - 4 | 508 | 68 | ||
| GenBank mRNA | DQ640760.1 | - | 3 - 4 | 424 | 68 | |
| DQ640761.1 | - | 2 - 3 | 628 | 68 | ||
| DQ640762.1 | - | 3 - 4 | 424 | 68 | ||
| DQ640763.1 | - | 2 - 3 | 518 | 68 | ||
More Information
Set Membership: |
|

PRKAB2
Genomic Map