Assay Details
| Gene Symbol: CASP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044689.1 | NP_001038154.1 | 3 - 4 | 706 | 126 | |
| XM_025152263.1 | XP_025008031.1 | 3 - 4 | 528 | 126 | ||
| GenBank mRNA | DQ010055.1 | - | 3 - 4 | 706 | 126 | |
| EU165363.1 | - | 2 - 3 | 418 | 126 | ||
| EU165364.1 | - | 2 - 3 | 418 | 126 | ||
More Information
Set Membership: |
|

CASP18
Genomic Map