Assay Details
| Gene Symbol: CASP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044689.1 | NP_001038154.1 | 6 - 7 | 1035 | 137 | |
| XM_025152263.1 | XP_025008031.1 | 6 - 7 | 857 | 137 | ||
| GenBank mRNA | DQ010055.1 | - | 6 - 7 | 1035 | 137 | |
| EU165363.1 | - | 5 - 6 | 747 | 137 | ||
More Information
Set Membership: |
|

CASP18
Genomic Map