Assay Details
| Gene Symbol: STK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045833.1 | NP_001039298.1 | 4 - 5 | 915 | 71 | |
| XM_015299932.2 | XP_015155418.1 | 4 - 5 | 997 | 71 | ||
| GenBank mRNA | DQ864976.1 | - | 4 - 5 | 915 | 71 | |
More Information
Set Membership: |
|

STK11
Genomic Map