Assay Details
| Gene Symbol: ZPAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045837.1 | NP_001039302.1 | 10 - 11 | 1368 | 149 | |
| XM_015284957.1 | XP_015140443.1 | 12 - 13 | 1537 | 149 | ||
More Information
Set Membership: |
|

ZPAX
Genomic Map