Assay Details
| Gene Symbol: ZPAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045837.1 | NP_001039302.1 | 2 - 3 | 171 | 149 | |
| XM_015284957.1 | XP_015140443.1 | 4 - 5 | 340 | 149 | ||
| GenBank mRNA | BX933361.1 | - | 4 - 5 | 347 | 149 | |
More Information
Set Membership: |
|

ZPAX
Genomic Map