Assay Details
| Gene Symbol: ZPAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045837.1 | NP_001039302.1 | 7 - 8 | 889 | 93 | |
| XM_015284957.1 | XP_015140443.1 | 9 - 10 | 1058 | 93 | ||
More Information
Set Membership: |
|

ZPAX
Genomic Map