Assay Details
| Gene Symbol: VIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048076.2 | NP_001041541.2 | 7 - 8 | 1321 | 69 | |
| GenBank mRNA | BU132443.1 | - | 7 - 8 | 743 | 69 | |
| BU297267.1 | - | 2 - 3 | 146 | 69 | ||
| BU354329.1 | - | 1 - 2 | 37 | 69 | ||
More Information
Set Membership: |
|

VIM
Genomic Map