Assay Details
| Gene Symbol: CX3CL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077232.1 | NP_001070700.1 | 2 - 3 | 214 | 96 | |
| GenBank mRNA | AM398231.1 | - | 2 - 3 | 214 | 96 | |
More Information
Set Membership: |
|

CX3CL1
Genomic Map