Assay Details
| Gene Symbol: CYCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079478.1 | NP_001072946.1 | 1 - 2 | 171 | 106 | |
| XM_015281451.2 | XP_015136937.1 | 2 - 3 | 316 | 106 | ||
| XM_015281452.2 | XP_015136938.1 | 2 - 3 | 307 | 106 | ||
| XM_015281453.2 | XP_015136939.1 | 2 - 3 | 286 | 106 | ||
More Information
Set Membership: |
|

CYCS
Genomic Map