Assay Details
| Gene Symbol: HMGN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079485.1 | NP_001072953.1 | 5 - 6 | 411 | 61 | |
| GenBank mRNA | BX950410.1 | - | 4 - 5 | 229 | 61 | |
| Y00416.1 | - | 411 | 61 | |||
More Information
Set Membership: |
|

HMGN4
Genomic Map