Assay Details
| Gene Symbol: DUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079488.1 | NP_001072956.1 | 1 - 2 | 170 | 81 | |
| XM_015291930.2 | XP_015147416.1 | 2 - 3 | 309 | 81 | ||
| XM_015291931.2 | XP_015147417.1 | 2 - 3 | 357 | 81 | ||
| XM_025153874.1 | XP_025009642.1 | 2 - 3 | 251 | 81 | ||
| GenBank mRNA | AJ720091.1 | - | 1 - 2 | 170 | 81 | |
More Information
Set Membership: |
|

DUT
Genomic Map