Assay Details
| Gene Symbol: RPL37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079493.2 | NP_001072961.1 | 2 - 3 | 147 | 61 | |
| GenBank mRNA | CD764696.1 | - | 2 - 3 | 147 | 61 | |
| CO504740.1 | - | 1 - 2 | 189 | 61 | ||
| D10731.1 | - | 1 - 2 | 68 | 61 | ||
More Information
Set Membership: |
|

RPL37A
Genomic Map