Assay Details
Gene Symbol: ZNF384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079496.1 | NP_001072964.1 | 8 - 9 | 1390 | 72 | |
XM_025151590.1 | XP_025007358.1 | 10 - 11 | 3052 | 72 | ||
XM_025151592.1 | XP_025007360.1 | 10 - 11 | 1471 | 72 | ||
XM_025151598.1 | XP_025007366.1 | 10 - 11 | 3082 | 72 | ||
XM_025151600.1 | XP_025007368.1 | 10 - 11 | 1512 | 72 | ||
XM_025151602.1 | XP_025007370.1 | 10 - 11 | 1499 | 72 | ||
XM_025151604.1 | XP_025007372.1 | 9 - 10 | 2896 | 72 | ||
XM_025151605.1 | XP_025007373.1 | 9 - 10 | 1317 | 72 | ||
XM_025151606.1 | XP_025007374.1 | 10 - 11 | 3056 | 72 | ||
XM_025151609.1 | XP_025007377.1 | 9 - 10 | 1356 | 72 | ||
XM_025151611.1 | XP_025007379.1 | 9 - 10 | 2892 | 72 | ||
XM_025151614.1 | XP_025007382.1 | 9 - 10 | 1312 | 72 | ||
XM_025151616.1 | XP_025007384.1 | 9 - 10 | 1352 | 72 | ||
XM_025151623.1 | XP_025007391.1 | 9 - 10 | 2970 | 72 | ||
XM_025151631.1 | XP_025007399.1 | 9 - 10 | 2869 | 72 | ||
XM_025151638.1 | XP_025007406.1 | 8 - 9 | 2786 | 72 | ||
GenBank mRNA | AJ719557.1 | - | 8 - 9 | 1390 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
