Assay Details
| Gene Symbol: WRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079497.1 | NP_001072965.1 | 4 - 5 | 494 | 62 | |
| XM_015300484.2 | XP_015155970.1 | 4 - 5 | 476 | 62 | ||
| GenBank mRNA | AJ720412.1 | - | 4 - 5 | 494 | 62 | |
| EU477532.1 | - | 3 - 4 | 306 | 62 | ||
| FJ882412.1 | - | 4 - 5 | 422 | 62 | ||
More Information
Set Membership: |
|

WRB
Genomic Map