Assay Details
| Gene Symbol: DENR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079505.1 | NP_001072973.1 | 4 - 5 | 322 | 99 | |
| XM_015275344.1 | XP_015130830.1 | 4 - 5 | 581 | 99 | ||
| XM_025155511.1 | XP_025011279.1 | 4 - 5 | 285 | 99 | ||
| GenBank mRNA | AJ720595.1 | - | 4 - 5 | 322 | 99 | |
| CR354249.1 | - | 2 - 3 | 494 | 99 | ||
More Information
Set Membership: |
|

DENR
Genomic Map