Assay Details
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079728.1 | NP_001073196.1 | 8 - 9 | 910 | 104 | |
XM_025149562.1 | XP_025005330.1 | 8 - 9 | 907 | 104 | ||
GenBank mRNA | AJ720025.1 | - | 8 - 9 | 910 | 104 | |
EU477531.1 | - | 7 - 8 | 857 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
