Assay Details
| Gene Symbol: LOC769174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079740.1 | NP_001073208.1 | 2 - 3 | 224 | 79 | |
| GenBank mRNA | DQ156496.1 | - | 2 - 3 | 224 | 79 | |
| DQ241793.1 | - | 2 - 3 | 224 | 79 | ||
More Information
Set Membership: |
|

LOC769174
Genomic Map